missing translation for 'onlineSavingsMsg'
Learn More

8-(2,3-Dichlorophenyl)-8-aza-5-azoniaspiro[4.5]decane Bromide, TRC

Product Code. 30380755
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380755 1 g -
30369565 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30380755 Supplier Toronto Research Chemicals Supplier No. TRCD4360701G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 795313-24-5
Molecular Formula C14 H19 Cl2 N2 . Br
Molecular Weight (g/mol) 366.12
Synonym 8-Aza-5-azoniaspiro[4.5]decane, 8-(2,3-dichlorophenyl)-, bromide (1:1), 8-Aza-5-azoniaspiro[4.5]decane, 8-(2,3-dichlorophenyl)-, bromide (9CI), 8-(2,3-Dichlorophenyl)-8-aza-5-azoniaspiro[4.5]decane bromide (1:1)
IUPAC Name 8-(2,3-dichlorophenyl)-8-aza-5-azoniaspiro[4.5]decane;bromide
SMILES [Br-].Clc1cccc(N2CC[N+]3(CCCC3)CC2)c1Cl

Specifications

Quantity 1 g
InChI Formula InChI=1S/C14H19Cl2N2.BrH/c15-12-4-3-5-13(14(12)16)17-6-10-18(11-7-17)8-1-2-9-18;/h3-5H,1-2,6-11H2;1H/q+1;/p-1
Formula Weight 364.0109
Chemical Name or Material 8-(2,3-Dichlorophenyl)-8-aza-5-azoniaspiro[4.5]decane Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.