CAS RN 795313-24-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
8-(2,3-Dichlorophenyl)-8-aza-5-azoniaspiro[4.5]decane Bromide, TRC
CAS: 795313-24-5 Molecular Formula: C14 H19 Cl2 N2 . Br Molecular Weight (g/mol): 366.12 Synonym: 8-Aza-5-azoniaspiro[4.5]decane, 8-(2,3-dichlorophenyl)-, bromide (1:1),8-Aza-5-azoniaspiro[4.5]decane, 8-(2,3-dichlorophenyl)-, bromide (9CI),8-(2,3-Dichlorophenyl)-8-aza-5-azoniaspiro[4.5]decane bromide (1:1) IUPAC Name: 8-(2,3-dichlorophenyl)-8-aza-5-azoniaspiro[4.5]decane;bromide SMILES: [Br-].Clc1cccc(N2CC[N+]3(CCCC3)CC2)c1Cl