missing translation for 'onlineSavingsMsg'
Learn More

8,14beta-Dihydrooripavine, TRC

Product Code. 30381573
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381573 25 mg 25mg
30374471 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30381573 Supplier Toronto Research Chemicals Supplier No. TRCD44996025MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873087-79-7
Molecular Formula C18 H21 N O3
Molecular Weight (g/mol) 299.36
Synonym Morphinan-3-ol, 6,7-didehydro-4,5-epoxy-6-methoxy-17-methyl-, (5α)-, (5α)-6,7-Didehydro-4,5-epoxy-6-methoxy-17-methylmorphinan-3-ol, 8,14-Dihydrooripavine, 8,14β-Dihydrooripavine
IUPAC Name (4R,4aR,7aR,12bS)-7-methoxy-3-methyl-2,4,4a,5,7a,13-hexahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-9-ol
SMILES COC1=CC[C@H]2[C@H]3Cc4ccc(O)c5O[C@@H]1[C@]2(CCN3C)c45

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C18H21NO3/c1-19-8-7-18-11-4-6-14(21-2)17(18)22-16-13(20)5-3-10(15(16)18)9-12(11)19/h3,5-6,11-12,17,20H,4,7-9H2,1-2H3/t11-,12+,17-,18-/m0/s1
Formula Weight 299.1521
Chemical Name or Material 8,14beta-Dihydrooripavine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.