CAS RN 873087-79-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
8,14beta-Dihydrooripavine, TRC
CAS: 873087-79-7 Molecular Formula: C18 H21 N O3 Molecular Weight (g/mol): 299.36 Synonym: Morphinan-3-ol, 6,7-didehydro-4,5-epoxy-6-methoxy-17-methyl-, (5α)-,(5α)-6,7-Didehydro-4,5-epoxy-6-methoxy-17-methylmorphinan-3-ol,8,14-Dihydrooripavine,8,14β-Dihydrooripavine IUPAC Name: (4R,4aR,7aR,12bS)-7-methoxy-3-methyl-2,4,4a,5,7a,13-hexahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-9-ol SMILES: COC1=CC[C@H]2[C@H]3Cc4ccc(O)c5O[C@@H]1[C@]2(CCN3C)c45