missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ 7X™-O-Matic Solution

7X™-O-Matic Solution was developed for safe and effective cleaning of laboratory instruments and glassware under pressure, heat and agitation (machine wash)

Brand:  MP Biomedicals™ 097667494

Code : NEW

Additional Details : Weight : 0.01000kg


 View more versions of this product

Product Code. 11449585

  • £314.82 / Pack of 4
Estimated Shipment: 19-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Developed for safe and effective cleaning of laboratory instruments and glassware
  • Formulated to be used in a machine washer to withstand heat, pressure and agitation
  • Powerful wetting and penetrating agent, which contains a non-ionic surfactant and a hydrotrope to complex phosphates
  • Behaves and remains mostly suds-free when agitated
  • Several convenient package sizes are available, and it is stable for long-term storage
  • Effective and safe
  • Eliminates residue leaving no interfering fluorescence
  • No etch to glass or plastic labware at any concentration
  • Completely drains from glassware in seconds
Specifications

Specifications

Liquid
Clear to Light Yellow
15°C to 30°C
4 Pk.
9 to 11
7X
Use on Glass, Plastic, benchtop equipment and tubing
1 gal.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 7X-O-Matic

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H318-Causes serious eye damage.

Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers