missing translation for 'onlineSavingsMsg'
Learn More

(7E,9E)-b-Ionylidene Acetaldehyde Technical grade (>85%), TRC

Product Code. 30395928
Change view
Click to view available options
Quantity:
25 mg
250 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395928 25 mg -
30402186 500 mg 500mg
30403900 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395928 Supplier Toronto Research Chemicals Supplier No. TRCI73400025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3917-41-7
Molecular Formula C15 H22 O
Molecular Weight (g/mol) 218.33
Synonym 2,4-Pentadienal, 3-methyl-5-(2,6,6-trimethyl-1-cyclohexen-1-yl)-, (2E,4E)-, 2,4-Pentadienal, 3-methyl-5-(2,6,6-trimethyl-1-cyclohexen-1-yl)-, (E,E)- (8CI), (2E,4E)-3-Methyl-5-(2,6,6-trimethyl-1-cyclohexen-1-yl)-2,4-pentadienal, (7E,9E)-β-Ionylideneacetaldehyde, (E)-β-Ionylideneacetaldehyde, (E,E)-3-Methyl-5-(2,6,6-trimethyl-1-cyclohexen-1-yl)-2,4-pentadienal, (2E,4E)-3-Methyl-5-(2,6,6-trimethylcyclohex-1-en-1-yl)penta-2,4-dienal
IUPAC Name (2E,4E)-3-methyl-5-(2,6,6-trimethylcyclohexen-1-yl)penta-2,4-dienal
SMILES C\C(=C/C=O)\C=C\C1=C(C)CCCC1(C)C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C15H22O/c1-12(9-11-16)7-8-14-13(2)6-5-10-15(14,3)4/h7-9,11H,5-6,10H2,1-4H3/b8-7+,12-9+
Formula Weight 218.1671
Chemical Name or Material (7E,9E)-beta-Ionylidene Acetaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.