CAS RN 3917-41-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(7E,9E)-b-Ionylidene Acetaldehyde Technical grade (>85%), TRC
CAS: 3917-41-7 Molecular Formula: C15 H22 O Molecular Weight (g/mol): 218.33 Synonym: 2,4-Pentadienal, 3-methyl-5-(2,6,6-trimethyl-1-cyclohexen-1-yl)-, (2E,4E)-,2,4-Pentadienal, 3-methyl-5-(2,6,6-trimethyl-1-cyclohexen-1-yl)-, (E,E)- (8CI),(2E,4E)-3-Methyl-5-(2,6,6-trimethyl-1-cyclohexen-1-yl)-2,4-pentadienal,(7E,9E)-β-Ionylideneacetaldehyde,(E)-β-Ionylideneacetaldehyde,(E,E)-3-Methyl-5-(2,6,6-trimethyl-1-cyclohexen-1-yl)-2,4-pentadienal,(2E,4E)-3-Methyl-5-(2,6,6-trimethylcyclohex-1-en-1-yl)penta-2,4-dienal IUPAC Name: (2E,4E)-3-methyl-5-(2,6,6-trimethylcyclohexen-1-yl)penta-2,4-dienal SMILES: C\C(=C/C=O)\C=C\C1=C(C)CCCC1(C)C