Learn More
Hamilton™ 700 Series Cemented Needle Custom Syringes
Syringes for customized needles. This series of syringes is ideal for dispensing volumes from 0.5 up to 500μL.
Brand:  Hamilton™ 87908
Code : NEW
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 10579672
  • £191.00 / Each
Estimated Shipment: 21-05-2024
to see stock.
Description
Description
  • The original hand-fitted Hamilton™ syringe
  • Stainless steel plunger is manufactured to fit the glass barrel with a tolerance less than three thousandths of a millimeter
  • Unsurpassed syringe life
Specifications
Specifications
Borosilicate Glass, Stainless Steel
Syringe
5 μL
6.6 mm
1/Ea.
Cemented Needle Special