Learn More
Hamilton™ 700 Series Cemented Needle Custom Syringes
Syringes for customized needles. This series of syringes is ideal for dispensing volumes from 0.5 up to 500μL.
£75.00 - £191.00
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10664301
|
Hamilton™
80308 |
10 μL |
£75.00
Each |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10359514
|
Hamilton™
80708 |
250 μL |
£143.00
Each |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10579672
|
Hamilton™
87908 |
5 μL |
£191.00
Each |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- The original hand-fitted Hamilton™ syringe
- Stainless steel plunger is manufactured to fit the glass barrel with a tolerance less than three thousandths of a millimeter
- Unsurpassed syringe life
Order Info
This is a Custom Order. Please consult with your Fisher Sales Representative before ordering.
Specifications
1/Ea. |