missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Ketocholesterol Antibody (7E1), Novus Biologicals™
Mouse Monoclonal Antibody
£150.00 - £484.00
Specifications
| Antigen | 7-Ketocholesterol |
|---|---|
| Clone | 70 |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18247221
|
Novus Biologicals
NBP2-59356 |
100 μg |
£484.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18656647
|
Novus Biologicals
NBP2-59356-25ug |
25 μg |
£150.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
7-Ketocholesterol Monoclonal specifically detects 7-Ketocholesterol in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| 7-Ketocholesterol | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Synthetic 7-Ketocholesterol conjugated to Keyhole Limpet Hemocyanin (KLH). | |
| Primary | |
| Specific for 7-ketocholesterol modified proteins. |
| 70 | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| 7KC, 7-KC, 7KC-Modified KLH | |
| IgG2b | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title