missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Ketocholesterol Antibody (7E1), Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP2-59356
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
7-Ketocholesterol Monoclonal specifically detects 7-Ketocholesterol in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| 7-Ketocholesterol | |
| Monoclonal | |
| Unconjugated | |
| PBS pH 7.4, 50% glycerol with 0.09% Sodium Azide | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Specific for 7-ketocholesterol modified proteins. | |
| Store at -20C. | |
| IgG2b |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 70 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| 7KC, 7-KC, 7KC-Modified KLH | |
| Synthetic 7-Ketocholesterol conjugated to Keyhole Limpet Hemocyanin (KLH). | |
| 100 μg | |
| Primary | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction