missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxyflavanone, 98%, Thermo Scientific™

Product Code. 10314452 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10314452 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10314452 Supplier Thermo Scientific Supplier No. 364170010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 6515-36-2
CAS Min % 97.5
CAS Max % 100.0
Melting Point 189°C to 197°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C15H12O3
MDL Number MFCD00017487
Quantity 1 g
Beilstein 18, V,2, 189
Synonym 7-hydroxyflavanone, 7-hydroxy-2-phenylchroman-4-one, chembl97542, 7-hydroxy-2-phenyl-4-chromanone, 7-hydroxy-2-phenyl-2,3-dihydro-4h-chromen-4-one, 2,3-dihydro-7-hydroxy-2-phenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 2,3-dihydro-7-hydroxy-2-phenyl, 7-hydroxy-flavanone, d05zpo
InChI Key SWAJPHCXKPCPQZ-UHFFFAOYSA-N
SMILES C1C(OC2=C(C1=O)C=CC(=C2)O)C3=CC=CC=C3
IUPAC Name 7-hydroxy-2-phenyl-2,3-dihydrochromen-4-one
Molecular Weight (g/mol) 240.26
PubChem CID 1890
ChEBI CHEBI:34483
Formula Weight 240.26
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 7-Hydroxyflavanone, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 7-Hydroxyflavanone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.