missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy-N-des{[2-(2-hydroxy)ethoxy]ethyl} Quetiapine Dihydrochloride, TRC

Product Code. 30392872
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392872 10 mg -
30407762 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30392872 Supplier Toronto Research Chemicals Supplier No. TRCH93610010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1397181-83-7
Molecular Formula C17 H17 N3 O S . 2 Cl H
Molecular Weight (g/mol) 384.32
Synonym Dibenzo[b,f][1,4]thiazepin-7-ol, 11-(1-piperazinyl)-, hydrochloride (1:2), 11-(Piperazin-1-yl)dibenzo[b,f][1,4]thiazepin-7-ol dihydrochloride, 7-Hydroxy-N-des{[2-(2-hydroxy)ethoxy]ethyl} Quetiapine Dihydrochloride
IUPAC Name 6-piperazin-1-ylbenzo[b][1,4]benzothiazepin-2-ol;dihydrochloride
SMILES Cl.Cl.Oc1ccc2N=C(N3CCNCC3)c4ccccc4Sc2c1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C17H17N3OS.2ClH/c21-12-5-6-14-16(11-12)22-15-4-2-1-3-13(15)17(19-14)20-9-7-18-8-10-20;;/h1-6,11,18,21H,7-10H2;2*1H
Formula Weight 383.0626
Chemical Name or Material 11-(Piperazin-1-yl)dibenzo[b,f][1,4]thiazepin-7-ol dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.