CAS RN 1397181-83-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
7-Hydroxy-N-des{[2-(2-hydroxy)ethoxy]ethyl} Quetiapine Dihydrochloride, TRC
CAS: 1397181-83-7 Molecular Formula: C17 H17 N3 O S . 2 Cl H Molecular Weight (g/mol): 384.32 Synonym: Dibenzo[b,f][1,4]thiazepin-7-ol, 11-(1-piperazinyl)-, hydrochloride (1:2),11-(Piperazin-1-yl)dibenzo[b,f][1,4]thiazepin-7-ol dihydrochloride,7-Hydroxy-N-des{[2-(2-hydroxy)ethoxy]ethyl} Quetiapine Dihydrochloride IUPAC Name: 6-piperazin-1-ylbenzo[b][1,4]benzothiazepin-2-ol;dihydrochloride SMILES: Cl.Cl.Oc1ccc2N=C(N3CCNCC3)c4ccccc4Sc2c1