missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy Coumarin-d5 Sulfate Potassium Salt, TRC

Product Code. 30403905
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30403905 2.5 mg 2.5mg
30399628 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30403905 Supplier Toronto Research Chemicals Supplier No. TRCH9248932.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1261392-49-7
Molecular Formula C9D5KO6S
Molecular Weight (g/mol) 285.33
Synonym 7-Sulfooxy-2H-1-benzopyran-2-one-d5 Potassium Salt, Umbelliferone-d5 Sulfate Potassium Salt
SMILES O=C1OC2=C([2H])C(OS(=O)(O[K])=O)=C([2H])C([2H])=C2C([2H])=C1[2H]

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C9H6O6S.K/c10-9-4-2-6-1-3-7(5-8(6)14-9)15-16(11,12)13;/h1-5H,(H,11,12,13);/q;+1/p-1/i1D,2D,3D,4D,5D;
Formula Weight 284.9758
Chemical Name or Material 7-Hydroxy Coumarin-d5 Sulfate Potassium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.