CAS RN 1261392-49-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
7-Hydroxy Coumarin-d5 Sulfate Potassium Salt, TRC
CAS: 1261392-49-7 Molecular Formula: C9D5KO6S Molecular Weight (g/mol): 285.33 Synonym: 7-Sulfooxy-2H-1-benzopyran-2-one-d5 Potassium Salt,Umbelliferone-d5 Sulfate Potassium Salt SMILES: O=C1OC2=C([2H])C(OS(=O)(O[K])=O)=C([2H])C([2H])=C2C([2H])=C1[2H]