missing translation for 'onlineSavingsMsg'
Learn More

7-Ethyltryptophol, TRC

Product Code. 30373924
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30373924 1 g
30382984 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30373924 Supplier Toronto Research Chemicals Supplier No. TRCE9318801G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41340-36-7
Molecular Formula C12 H15 N O
Molecular Weight (g/mol) 189.25
Synonym 2-(7-Ethylindol-3-yl)ethanol, 1H-Indole-3-ethanol, 7-ethyl-, 7-Ethyl-1H-indole-3-ethanol, 2-(7-Ethyl-1H-indol-3-yl)-ethanol, 3-(2-Hydroxyethyl)-7-ethylindole, 7-Ethyl-3-indoleethanol, 7-Ethyltryptophol, RAK 801
IUPAC Name 2-(7-ethyl-1H-indol-3-yl)ethanol
SMILES CCc1cccc2c(CCO)c[nH]c12

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H15NO/c1-2-9-4-3-5-11-10(6-7-14)8-13-12(9)11/h3-5,8,13-14H,2,6-7H2,1H3
Formula Weight 189.1154
Chemical Name or Material 2-(7-Ethylindol-3-yl)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.