accessibility menu, dialog, popup

UserName

CH3HOHN

CAS RN 41340-36-7

IUPAC Name: 2-(7-ethyl-1H-indol-3-yl)ethan-1-ol
Synonyms: 2-(7-ethyl-1H-indol-3-yl)ethanol
Molecular Weight (g/mol): 189.26
Molecular Formula: C12H15NO
InChi Key: UVSDNCAZVSQJQA-UHFFFAOYSA-N
SMILES: CCC1=C2NC=C(CCO)C2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results