missing translation for 'onlineSavingsMsg'
Learn More

7-Bromoisoquinoline N-oxide, 98%, Thermo Scientific Chemicals

Product Code. 11465650
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11465650 25 g 25g
11455650 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11465650 Supplier Thermo Scientific Alfa Aesar Supplier No. H25899.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

7-Bromoisoquinoline N-oxide is the reactant in the direct regioselective phosphonation of heteroaryl N-oxides with H-phosphonates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Bromoisoquinoline N-oxide is the reactant in the direct regioselective phosphonation of heteroaryl N-oxides with H-phosphonates

Solubility
Soluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 223671-18-9
Molecular Formula C9H6BrNO
Molecular Weight (g/mol) 224.057
MDL Number MFCD09953462
InChI Key QLRDQGRZLHHBPE-UHFFFAOYSA-N
Synonym 7-bromoisoquinolin-2-ium-2-olate, 7-bromoisoquinoline-n-oxide, 7-bromoisoquinoline 2-oxide, 7-bromoisoquinoline n-oxide
PubChem CID 22665927
IUPAC Name 7-bromo-2-oxidoisoquinolin-2-ium
SMILES C1=CC(=CC2=C1C=C[N+](=C2)[O-])Br

Specifications

Melting Point 181°C to 188°C
Quantity 25 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 224.06
Percent Purity 98%
Chemical Name or Material 7-Bromoisoquinoline N-oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.