accessibility menu, dialog, popup

UserName

NOBr

CAS RN 223671-18-9

IUPAC Name: 7-bromoisoquinolin-2-ium-2-olate
Synonyms: 7-bromoisoquinolin-2-ium-2-olate
Molecular Weight (g/mol): 224.06
Molecular Formula: C9H6BrNO
InChi Key: QLRDQGRZLHHBPE-UHFFFAOYSA-N
SMILES: [O-][N+]1=CC2=CC(Br)=CC=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results