missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ 7-AAD Viability Staining Solution

Product Code. 15239004
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15239004 2 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Contains Azide
Product Code. 15239004 Supplier Invitrogen™ Supplier No. 00699350
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
Contains Azide

Ready-to-use solution for the exclusion of nonviable cells in flow cytometric analysis. Can be used in combination with PE- and FITC-conjugated antibodies in 2-color analysis. The advantage of 7-AAD over PI is that there is minimal spectral overlap between these emissions.

Fluorescence is detected in the far red range of the spectrum (650nm long-pass filter)
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Flow Cytometer
Product Type Viability Staining Solution
Dye Type Cell-Impermeant
Emission 535/617
Product Line eBioscience
Quantity 2 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.