missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies 7-AAD (7-Aminoactinomycin D)

Product Code. 17170352
Encompass_Preferred
Click to view available options
Quantity:
200 Tests
500 Tests
Unit Size:
200 tests
500 tests
This item is not returnable. View return policy

Product Code. 17170352

Brand: STEMCELL Technologies 75001.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cell viability dye (DNA-labeling dye)

  • 7‐AAD (7‐Aminoactinomycin D) is a fluorescent cell viability dye which is excluded from live cells with intact membranes but penetrates dead or damaged cells and binds to double‐stranded DNA with high affinity by intercalating between GC base pairs
  • It is often used as an alternative to propidium iodide (PI; Catalog #75002) to differentiate and exclude non-viable cells in flow cytometric analyses, and can be excited using 488 nm (blue), 532 nm (green), and 561 nm (yellow-green) laser lines
  • Compared to PI, 7-AAD offers the advantage of having minimal overlap in its emission spectra with mostly blue and green, and many red fluorophores, including the commonly used phycoerythrin (PE) and fluorescein isothiocyanate (FITC)
  • 7‐AAD is also used in DNA fluorescence imaging applications to discriminate between early and late stages of apoptosis, to study cell‐mediated cytotoxicity

Specifications

Quantity 200 Tests
For Use With (Application) Flow Cytometry, Immunocytochemistry, Immunofluorescence
Cell Type Other
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.