missing translation for 'onlineSavingsMsg'
Learn More

7-(4-Iodobutoxy)-3,4-dihydroquinolin-2-one, TRC

Product Code. 30137576
Change view
Click to view available options
Quantity:
25 mg
100 mg
500 mg
Unit Size:
100mg
25mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137576 500 mg 500mg
30134449 100 mg 100mg
17953895 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137576 Supplier Toronto Research Chemicals Supplier No. TRCI694200500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 952308-47-3
Molecular Formula C13 H16 I N O2
Molecular Weight (g/mol) 345.18
Synonym 2(1H)-Quinolinone, 3,4-dihydro-7-(4-iodobutoxy)-, 3,4-Dihydro-7-(4-iodobutoxy)-2(1H)-quinolinone, 7-(4-Iodobutoxy)-3,4-dihydro-2(1H)-quinolinone
IUPAC Name 7-(4-iodobutoxy)-3,4-dihydro-1H-quinolin-2-one
SMILES ICCCCOc1ccc2CCC(=O)Nc2c1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C13H16INO2/c14-7-1-2-8-17-11-5-3-10-4-6-13(16)15-12(10)9-11/h3,5,9H,1-2,4,6-8H2,(H,15,16)
Formula Weight 345.0226
Percent Purity >95%
Chemical Name or Material 7-(4-Iodobutoxy)-3,4-dihydroquinolin-2-one
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.