CAS RN 952308-47-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
7-(4-Iodobutoxy)-3,4-dihydro-2(1H)-quinolinone, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
7-(4-Iodobutoxy)-3,4-dihydroquinolin-2-one, TRC
CAS: 952308-47-3 Molecular Formula: C13 H16 I N O2 Molecular Weight (g/mol): 345.18 Synonym: 2(1H)-Quinolinone, 3,4-dihydro-7-(4-iodobutoxy)-,3,4-Dihydro-7-(4-iodobutoxy)-2(1H)-quinolinone,7-(4-Iodobutoxy)-3,4-dihydro-2(1H)-quinolinone IUPAC Name: 7-(4-iodobutoxy)-3,4-dihydro-1H-quinolin-2-one SMILES: ICCCCOc1ccc2CCC(=O)Nc2c1