missing translation for 'onlineSavingsMsg'
Learn More

7-(4-Bromobutoxy)-3,4-dihydroquinolin-2-one, TRC

Product Code. 30135125
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135125 5 g 5g
30134227 10 g 10g
30008719 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135125 Supplier Toronto Research Chemicals Supplier No. TRCB6819605G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129722-34-5
Molecular Formula C13 H16 Br N O2
Molecular Weight (g/mol) 298.18
Synonym 2(1H)-Quinolinone, 7-(4-bromobutoxy)-3,4-dihydro-, 7-(4-Bromobutoxy)-3,4-dihydro-2(1H)-quinolinone, 7-(4-Bromobutoxy)-3,4-dihydro-2-quinolinone
IUPAC Name 7-(4-bromobutoxy)-3,4-dihydro-1H-quinolin-2-one
SMILES BrCCCCOc1ccc2CCC(=O)Nc2c1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C13H16BrNO2/c14-7-1-2-8-17-11-5-3-10-4-6-13(16)15-12(10)9-11/h3,5,9H,1-2,4,6-8H2,(H,15,16)
Formula Weight 297.0364
Percent Purity >95%
Chemical Name or Material 7-(4-Bromobutoxy)-3,4-dihydroquinolin-2-one
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.