CAS RN 129722-34-5
CAS RN 129722-34-5
IUPAC Name:
7-(4-bromobutoxy)-1,2,3,4-tetrahydroquinolin-2-one
Synonyms:
7-(4-bromobutoxy)-3,4-dihydro-1H-quinolin-2-one
Molecular Weight (g/mol):
298.18
Molecular Formula:
C13H16BrNO2
InChi Key:
URHLNHVYMNBPEO-UHFFFAOYSA-N
SMILES:
BrCCCCOC1=CC2=C(CCC(=O)N2)C=C1
1
–
1
of
1
results
1
7-(4-Bromobutoxy)-3,4-dihydro-2(1H)-quinolinone, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.