missing translation for 'onlineSavingsMsg'
Learn More

6alpha,9alpha-Difluoroprednisolone, TRC

Product Code. 30380331
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380331 250 mg
30374588 50 mg
30388537 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380331 Supplier Toronto Research Chemicals Supplier No. TRCD446290250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 806-29-1
Molecular Formula C21 H26 F2 O5
Molecular Weight (g/mol) 396.42
Synonym 6α,9-difluoro-11β,17,21-trihydroxy-pregna-1,4-diene-3,20-dione, 6-alpha-Fluoro-isoflupredone, 6α,9-Difluoroprednisolone, NSC 77021
IUPAC Name (6S,8S,9R,10S,11S,13S,14S,17R)-6,9-difluoro-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
SMILES C[C@]12C[C@H](O)[C@@]3(F)[C@@H](C[C@H](F)C4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C21H26F2O5/c1-18-5-3-11(25)7-14(18)15(22)8-13-12-4-6-20(28,17(27)10-24)19(12,2)9-16(26)21(13,18)23/h3,5,7,12-13,15-16,24,26,28H,4,6,8-10H2,1-2H3/t12-,13-,15-,16-,18-,19-,20-,21-/m0/s1
Formula Weight 396.1748
Chemical Name or Material 6alpha,9alpha-Difluoroprednisolone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.