CAS RN 806-29-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6alpha,9alpha-Difluoroprednisolone, TRC
CAS: 806-29-1 Molecular Formula: C21 H26 F2 O5 Molecular Weight (g/mol): 396.42 Synonym: 6α,9-difluoro-11β,17,21-trihydroxy-pregna-1,4-diene-3,20-dione,6-alpha-Fluoro-isoflupredone,6α,9-Difluoroprednisolone,NSC 77021 IUPAC Name: (6S,8S,9R,10S,11S,13S,14S,17R)-6,9-difluoro-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one SMILES: C[C@]12C[C@H](O)[C@@]3(F)[C@@H](C[C@H](F)C4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO