missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agilent BioTek 615/10 nm Lanthascreen Europium Donor Emission
Shop All Agilent BioTek Products
Click to view available options
Wavelength:
615 ±10 nm
Unit Size:
Each
Description
- Accessory for Agilent BioTek instruments range of microplate washers, readers and cell imagers
- Fluorescence filter
- For use with single and multimode microplate readers and imagers
Specifications
Specifications
| For Use With (Equipment) | Microplate Readers and Imagers |
| Wavelength | 615 ±10 nm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction