missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-5-nitroquinoline, 97%, Thermo Scientific™

Product Code. p-7043444
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 11370234

Brand: Thermo Scientific Alfa Aesar L00807.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23141-61-9
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.186
MDL Number MFCD00024021
InChI Key CENBTULRDDPOGR-UHFFFAOYSA-N
Synonym 5-nitro-6-methylquinoline, quinoline, 6-methyl-5-nitro, quinoline,6-methyl-5-nitro, acmc-1cuis, maybridge1_002137, 5-nitro-6-met hylquinoline, 5-nitro-6-methyl quinoline, 6-methyl-5-nitroquinoline
PubChem CID 90009
IUPAC Name 6-methyl-5-nitroquinoline
SMILES CC1=C(C2=C(C=C1)N=CC=C2)[N+](=O)[O-]

Specifications

Melting Point 115°C to 118°C
Quantity 5 g
Beilstein 154688
Formula Weight 188.19
Percent Purity 97%
Chemical Name or Material 6-Methyl-5-nitroquinoline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.