accessibility menu, dialog, popup

UserName

CAS RN 23141-61-9

H3CNNOO

CAS RN 23141-61-9

IUPAC Name: 6-methyl-5-nitroquinoline
Synonyms: 6-methyl-5-nitroquinoline
Molecular Weight (g/mol): 188.19
Molecular Formula: C10H8N2O2
InChi Key: CENBTULRDDPOGR-UHFFFAOYSA-N
SMILES: CC1=CC=C2N=CC=CC2=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results