missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy Bexarotene, TRC

Product Code. 30394854
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394854 10 mg -
30402870 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30394854 Supplier Toronto Research Chemicals Supplier No. TRCH81750010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 368451-07-4
Molecular Formula C24 H28 O3
Molecular Weight (g/mol) 364.48
Synonym 4-[1-(5,6,7,8-Tetrahydro-6-hydroxy-3,5,5,8,8-pentamethyl-2-naphthalenyl)ethenyl]benzoic Acid
IUPAC Name 4-[1-(6-hydroxy-3,5,5,8,8-pentamethyl-6,7-dihydronaphthalen-2-yl)ethenyl]benzoic acid
SMILES Cc1cc2c(cc1C(=C)c3ccc(cc3)C(=O)O)C(C)(C)CC(O)C2(C)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C24H28O3/c1-14-11-20-19(23(3,4)13-21(25)24(20,5)6)12-18(14)15(2)16-7-9-17(10-8-16)22(26)27/h7-12,21,25H,2,13H2,1,3-6H3,(H,26,27)
Formula Weight 364.2038
Chemical Name or Material 6-Hydroxy Bexarotene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.