CAS RN 368451-07-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6-Hydroxy Bexarotene, TRC
CAS: 368451-07-4 Molecular Formula: C24 H28 O3 Molecular Weight (g/mol): 364.48 Synonym: 4-[1-(5,6,7,8-Tetrahydro-6-hydroxy-3,5,5,8,8-pentamethyl-2-naphthalenyl)ethenyl]benzoic Acid IUPAC Name: 4-[1-(6-hydroxy-3,5,5,8,8-pentamethyl-6,7-dihydronaphthalen-2-yl)ethenyl]benzoic acid SMILES: Cc1cc2c(cc1C(=C)c3ccc(cc3)C(=O)O)C(C)(C)CC(O)C2(C)C