missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy-2-naphthaleneacetic Acid, TRC

Product Code. 30012547
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012547 250 mg 250mg
30010353 50 mg 50mg
30010965 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012547 Supplier Toronto Research Chemicals Supplier No. TRCH948520250MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10441-46-0
Molecular Formula C12 H10 O3
Molecular Weight (g/mol) 202.21
IUPAC Name 2-(6-hydroxynaphthalen-2-yl)acetic acid
SMILES OC(=O)Cc1ccc2cc(O)ccc2c1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H10O3/c13-11-4-3-9-5-8(6-12(14)15)1-2-10(9)7-11/h1-5,7,13H,6H2,(H,14,15)
Formula Weight 202.063
Chemical Name or Material 6-Hydroxy-2-naphthaleneacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.