CAS RN 10441-46-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6-Hydroxy-2-naphthaleneacetic Acid, TRC
CAS: 10441-46-0 Molecular Formula: C12 H10 O3 Molecular Weight (g/mol): 202.21 IUPAC Name: 2-(6-hydroxynaphthalen-2-yl)acetic acid SMILES: OC(=O)Cc1ccc2cc(O)ccc2c1