missing translation for 'onlineSavingsMsg'
Learn More

6-(Gamma,Gamma-Dimethylallylamino)purine Arabinoside, TRC

Product Code. 30385416
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385416 50 mg
30387539 250 mg
30382556 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385416 Supplier Toronto Research Chemicals Supplier No. TRCD47413850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33156-15-9
Molecular Formula C15H21N5O4
Molecular Weight (g/mol) 335.36
Synonym 9-beta-D-Arabinofuranosyl-N-(3-methyl-2-butenyl)-9H-purin-6-amine, 9-beta-D-Arabinofuranosyl-N-(3-methyl-2-butenyl)-adenine, 9-beta-D-Arabinofuranosyl-N-(3-methyl-2-buten-1-yl)-9H-purin-6-amine
SMILES CC(=CCNc1ncnc2c1ncn2[C@@H]3O[C@H](CO)[C@@H](O)[C@@H]3O)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C15H21N5O4/c1-8(2)3-4-16-13-10-14(18-6-17-13)20(7-19-10)15-12(23)11(22)9(5-21)24-15/h3,6-7,9,11-12,15,21-23H,4-5H2,1-2H3,(H,16,17,18)/t9-,11-,12+,15-/m1/s1
Formula Weight 335.1594
Chemical Name or Material 6-(gamma,gamma-Dimethylallylamino)purine Arabinoside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.