accessibility menu, dialog, popup

UserName

CH3CH3HNNNNN(R)O(R)OH(S)HO(R)HO

CAS RN 33156-15-9

IUPAC Name: (2R,3S,4R,5R)-2-(hydroxymethyl)-5-{6-[(3-methylbut-2-en-1-yl)amino]-9H-purin-9-yl}oxolane-3,4-diol
Synonyms: isopentenyladenosine dimethylallyladenosine 2iPA N6-(2-isopentenyl)adenosine N6-isopentenyladenosine N(sup 6)-isopentenyladenosine isopentenyl adenine riboside riboprina riboprine i6a
Molecular Weight (g/mol): 335.36
Molecular Formula: C15H21N5O4
InChi Key: USVMJSALORZVDV-SDBHATRESA-N
SMILES: CC(C)=CCNC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)