missing translation for 'onlineSavingsMsg'
Learn More

6-Deoxypenciclovir, TRC

Product Code. 30137074
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137074 25 mg 25mg
30135778 100 mg 100mg
17961585 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137074 Supplier Toronto Research Chemicals Supplier No. TRCD24765025MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104227-86-3
Molecular Formula C10 H15 N5 O2
Molecular Weight (g/mol) 237.26
IUPAC Name 2-[2-(2-aminopurin-9-yl)ethyl]propane-1,3-diol
SMILES Nc1ncc2ncn(CCC(CO)CO)c2n1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C10H15N5O2/c11-10-12-3-8-9(14-10)15(6-13-8)2-1-7(4-16)5-17/h3,6-7,16-17H,1-2,4-5H2,(H2,11,12,14)
Formula Weight 237.1226
Percent Purity >95%
Chemical Name or Material 6-Deoxypenciclovir
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.