CAS RN 104227-86-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6-Deoxypenciclovir, TRC
CAS: 104227-86-3 Molecular Formula: C10 H15 N5 O2 Molecular Weight (g/mol): 237.26 IUPAC Name: 2-[2-(2-aminopurin-9-yl)ethyl]propane-1,3-diol SMILES: Nc1ncc2ncn(CCC(CO)CO)c2n1