missing translation for 'onlineSavingsMsg'
Learn More

6-Cyano-2-(2-pyridyl)benzimidazole, 97%, Thermo Scientific™

Product Code. p-7053004
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15474858 1 g 1g
15484858 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15474858 Supplier Thermo Scientific Alfa Aesar Supplier No. H58615.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 226702-84-7
Molecular Formula C13H8N4
Molecular Weight (g/mol) 220.235
MDL Number MFCD22581244
InChI Key JOPNJLSDCGCEFK-UHFFFAOYSA-N
Synonym 2-pyridin-2-yl-1h-1,3-benzodiazole-5-carbonitrile, 2-pyridin-2-yl-3h-1,3-benzodiazole-5-carbonitrile, 6-cyano-2-2-pyridyl benzimidazole, 2-pyridin-2-yl-1h-benzimidazole-5-carbonitrile, 2-pyridin-2-yl-1h-benzoimidazole-5-carbonitrile, 2-pyridin-2-yl-1h-benzo d imidazole-5-carbonitrile
PubChem CID 11543088
IUPAC Name 2-pyridin-2-yl-3H-benzimidazole-5-carbonitrile
SMILES C1=CC=NC(=C1)C2=NC3=C(N2)C=C(C=C3)C#N

Specifications

Color Brown
Quantity 1 g
UN Number UN3439
Formula Weight 220.23
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 6-Cyano-2-(2-pyridyl)benzimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.