accessibility menu, dialog, popup

UserName

NNHNN

CAS RN 226702-84-7

IUPAC Name: 2-(pyridin-2-yl)-1H-1,3-benzodiazole-6-carbonitrile
Synonyms: 2-(pyridin-2-yl)-3H-1,3-benzodiazole-5-carbonitrile
Molecular Weight (g/mol): 220.24
Molecular Formula: C13H8N4
InChi Key: JOPNJLSDCGCEFK-UHFFFAOYSA-N
SMILES: N#CC1=CC=C2N=C(NC2=C1)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results