missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-3-pyridineacetic acid, 97%

Product Code. 11357549
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357549 10 g 10g
11367549 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357549 Supplier Thermo Scientific Alfa Aesar Supplier No. H27966.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 39891-13-9 | C7H6ClNO2 | 171.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39891-13-9
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD01863172
InChI Key XWVPSJPQWOVRHJ-UHFFFAOYSA-N
Synonym 2-6-chloropyridin-3-yl acetic acid, 2-chloropyridine-5-acetic acid, 6-chloro-pyridin-3-yl-acetic acid, 6-chloro-3-pyridineacetic acid, 6-chloropyridin-3-yl acetic acid, 2-6-chloro-3-pyridyl acetic acid, 3-pyridineacetic acid, 6-chloro, 2-chloropyridyl-5-acetic acid, 2-2-chloropyridin-5-yl acetic acid, 6-chloropyridine-3-acetic acid
PubChem CID 14071571
IUPAC Name 2-(6-chloropyridin-3-yl)acetic acid
SMILES C1=CC(=NC=C1CC(=O)O)Cl

Specifications

Melting Point 172°C to 176°C
Quantity 10 g
Formula Weight 171.58
Percent Purity 97%
Chemical Name or Material 6-Chloro-3-pyridineacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.