CAS RN 39891-13-9
IUPAC Name:
2-(6-chloropyridin-3-yl)acetic acid
Synonyms:
(6-chloropyridin-3-yl)acetic acid
Molecular Weight (g/mol):
171.58
Molecular Formula:
C7H6ClNO2
InChi Key:
XWVPSJPQWOVRHJ-UHFFFAOYSA-N
SMILES:
OC(=O)CC1=CC=C(Cl)N=C1
1
–
1
of
1
results
1
6-Chloro-3-pyridineacetic acid, 97%
CAS: 39891-13-9 Molecular Formula: C7H6ClNO2 Molecular Weight (g/mol): 171.58 MDL Number: MFCD01863172 InChI Key: XWVPSJPQWOVRHJ-UHFFFAOYSA-N Synonym: 2-6-chloropyridin-3-yl acetic acid,2-chloropyridine-5-acetic acid,6-chloro-pyridin-3-yl-acetic acid,6-chloro-3-pyridineacetic acid,6-chloropyridin-3-yl acetic acid,2-6-chloro-3-pyridyl acetic acid,3-pyridineacetic acid, 6-chloro,2-chloropyridyl-5-acetic acid,2-2-chloropyridin-5-yl acetic acid,6-chloropyridine-3-acetic acid PubChem CID: 14071571 IUPAC Name: 2-(6-chloropyridin-3-yl)acetic acid SMILES: C1=CC(=NC=C1CC(=O)O)Cl