missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dihydro-9-hydroxy-6-oxo-5H-pyrido[3,2-b]azepine-8-carboxylic Acid Ethyl Ester, TRC

Product Code. 30376260
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30376260 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30376260 Supplier Toronto Research Chemicals Supplier No. TRCD449143100MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 676596-62-6
Molecular Formula C12 H12 N2 O4
Quantity 100 mg
InChI Formula InChI=1S/C12H12N2O4/c1-2-18-12(17)7-6-9(15)14-8-4-3-5-13-10(8)11(7)16/h3-5,16H,2,6H2,1H3,(H,14,15)
SMILES CCOC(=O)C1=C(O)c2ncccc2NC(=O)C1
IUPAC Name ethyl 9-hydroxy-6-oxo-5,7-dihydropyrido[3,2-b]azepine-8-carboxylate
Molecular Weight (g/mol) 248.23
Formula Weight 248.0797
Chemical Name or Material 6,7-Dihydro-9-hydroxy-6-oxo-5H-pyrido[3,2-b]azepine-8-carboxylic acid ethyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.