missing translation for 'onlineSavingsMsg'
Learn More

6-(1,4-Diazepan-1yl)nicotinonitrile, ≥97%, Thermo Scientific™

Product Code. 10694015
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10694015 10 g 10g
10061702 1 g 1g
10096414 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10694015 Supplier Thermo Scientific Maybridge Supplier No. SPB07871EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 683274-59-1
Molecular Formula C11H14N4
Molecular Weight (g/mol) 202.261
MDL Number MFCD01567269
InChI Key AWTOGESBXYCVMV-UHFFFAOYSA-N
Synonym 6-1,4-diazepan-1-yl nicotinonitrile, 6-1,4-diazepan-1-yl pyridine-3-carbonitrile, 3-pyridinecarbonitrile,6-hexahydro-1h-1,4-diazepin-1-yl, maybridge4_004605, 6-1,4-diazepan-1yl nicotinonitrile, 6-homopiperazin-1-yl nicotinonitrile, 6-1,4-diazepan-1-yl-3-pyridinecarbonitrile, 6-1,4-diazaperhydroepinyl pyridine-3-carbonitrile
PubChem CID 2747101
IUPAC Name 6-(1,4-diazepan-1-yl)pyridine-3-carbonitrile
SMILES C1CNCCN(C1)C2=NC=C(C=C2)C#N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 202.26
Percent Purity 97%
Chemical Name or Material 6-(1, 4-Diazepan-1-yl)nicotinonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.