CAS RN 683274-59-1
IUPAC Name:
6-(1,4-diazepan-1-yl)pyridine-3-carbonitrile
Synonyms:
6-(1,4-diazepan-1-yl)pyridine-3-carbonitrile
Molecular Weight (g/mol):
202.26
Molecular Formula:
C11H14N4
InChi Key:
AWTOGESBXYCVMV-UHFFFAOYSA-N
SMILES:
N#CC1=CN=C(C=C1)N1CCCNCC1
1
–
1
of
1
results
1
6-(1,4-Diazepan-1yl)nicotinonitrile, ≥97%, Thermo Scientific™
CAS: 683274-59-1 Molecular Formula: C11H14N4 Molecular Weight (g/mol): 202.261 MDL Number: MFCD01567269 InChI Key: AWTOGESBXYCVMV-UHFFFAOYSA-N Synonym: 6-1,4-diazepan-1-yl nicotinonitrile,6-1,4-diazepan-1-yl pyridine-3-carbonitrile,3-pyridinecarbonitrile,6-hexahydro-1h-1,4-diazepin-1-yl,maybridge4_004605,6-1,4-diazepan-1yl nicotinonitrile,6-homopiperazin-1-yl nicotinonitrile,6-1,4-diazepan-1-yl-3-pyridinecarbonitrile,6-1,4-diazaperhydroepinyl pyridine-3-carbonitrile PubChem CID: 2747101 IUPAC Name: 6-(1,4-diazepan-1-yl)pyridine-3-carbonitrile SMILES: C1CNCCN(C1)C2=NC=C(C=C2)C#N