missing translation for 'onlineSavingsMsg'
Learn More

6-(1,4-Diazepan-1yl)nicotinonitrile, ≥97%, Thermo Scientific™

Product Code. 10061702
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
This item is not returnable. View return policy

Product Code. 10061702

Brand: Thermo Scientific Maybridge SPB07871DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 683274-59-1
Molecular Formula C11H14N4
Molecular Weight (g/mol) 202.261
MDL Number MFCD01567269
InChI Key AWTOGESBXYCVMV-UHFFFAOYSA-N
Synonym 6-1,4-diazepan-1-yl nicotinonitrile, 6-1,4-diazepan-1-yl pyridine-3-carbonitrile, 3-pyridinecarbonitrile,6-hexahydro-1h-1,4-diazepin-1-yl, maybridge4_004605, 6-1,4-diazepan-1yl nicotinonitrile, 6-homopiperazin-1-yl nicotinonitrile, 6-1,4-diazepan-1-yl-3-pyridinecarbonitrile, 6-1,4-diazaperhydroepinyl pyridine-3-carbonitrile
PubChem CID 2747101
IUPAC Name 6-(1,4-diazepan-1-yl)pyridine-3-carbonitrile
SMILES C1CNCCN(C1)C2=NC=C(C=C2)C#N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 202.26
Percent Purity 97%
Chemical Name or Material 6-(1, 4-Diazepan-1-yl)nicotinonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.