missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5T4 Antibody (6E9), Novus Biologicals™
Rabbit Monoclonal Antibody
£269.00 - £406.00
Specifications
| Antigen | 5T4 |
|---|---|
| Clone | 6E9 |
| Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30029742
|
Novus Biologicals
NBP3-26276-50ul |
50 μL |
£269.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30032763
|
Novus Biologicals
NBP3-26276-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
5T4 Monoclonal antibody specifically detects 5T4 in Human, Mouse samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| 5T4 | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| 5T4 oncofetal trophoblast glycoprotein, 5T4 oncotrophoblast glycoprotein, 5T4AG, 5T4-AG, 5T4M6P1,5T4 oncofetal antigen, trophoblast glycoprotein | |
| A synthesized peptide derived from Human 5T4 [UniProt Q13641] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 6E9 | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Immunology, Stem Cells | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 7162 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title