missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5T4 Antibody (6E9), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26276-50ul
This item is not returnable.
View return policy
Description
5T4 Monoclonal antibody specifically detects 5T4 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| 5T4 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| 6E9 | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
| 5T4 oncofetal trophoblast glycoprotein, 5T4 oncotrophoblast glycoprotein, 5T4AG, 5T4-AG, 5T4M6P1,5T4 oncofetal antigen, trophoblast glycoprotein | |
| A synthesized peptide derived from Human 5T4 [UniProt Q13641] | |
| 50 μL | |
| Cancer, Cell Biology, Immunology, Stem Cells | |
| 7162 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction