missing translation for 'onlineSavingsMsg'
Learn More
Learn More
53BP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£248.00 - £458.00
Specifications
| Antigen | 53BP1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:5000-1:25000, Chromatin Immunoprecipitation reported in scientific literature (PMID 24591601), Flow Cytometry 1.5 ug/ml, Immunohistochemistry reported in scientific literature (PMID 24987917), Immunocytochemistry/Immunofluorescence 1:1000-1:5000, Immunoprecipitation reported in scientific literature (PMID 25645366), Immunohistochemistry-Paraffin 1:1000-1:5000. Use reported in scientific literature (PMID 27653664), Immunohistochemistry-Frozen reported by customer review, Immunoblotting reported in multiple pieces of scientific literature, In-situ Hybridization reported in scientific literature (PMID 34988401; 33122290), Flow (Intracellular), Chromatin Immunoprecipitation (ChIP), Knockout Validated reported in scientific literature (PMID 26601238), KnockDown Validated |
| Applications | Western Blot, ChIP Assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, In Situ Hybridization (ISH), Immunoassay, KnockDown |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18735403
|
Novus Biologicals
NB100-304SS |
0.025 mL |
£248.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18799923
|
Novus Biologicals
NB100-304 |
£458.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
53BP1 Polyclonal specifically detects 53BP1 in Human, Mouse, Rat, Bat, Bovine, Canine, Fish, Goat, Primate samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, In-situ Hybridization, Flow (Intracellular), Knockout Validated, Knockdown Validated.Specifications
| 53BP1 | |
| Western Blot 1:5000-1:25000, Chromatin Immunoprecipitation reported in scientific literature (PMID 24591601), Flow Cytometry 1.5 ug/ml, Immunohistochemistry reported in scientific literature (PMID 24987917), Immunocytochemistry/Immunofluorescence 1:1000-1:5000, Immunoprecipitation reported in scientific literature (PMID 25645366), Immunohistochemistry-Paraffin 1:1000-1:5000. Use reported in scientific literature (PMID 27653664), Immunohistochemistry-Frozen reported by customer review, Immunoblotting reported in multiple pieces of scientific literature, In-situ Hybridization reported in scientific literature (PMID 34988401; 33122290), Flow (Intracellular), Chromatin Immunoprecipitation (ChIP), Knockout Validated reported in scientific literature (PMID 26601238), KnockDown Validated | |
| Polyclonal | |
| Rabbit | |
| Breast Cancer, Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Tumor Suppressors | |
| PBS with 0.05% Sodium Azide | |
| 53BP1, p202, p53-binding protein 1, p53bp1, TDRD30, TP53-Binding Protein 1, tumor protein 53-binding protein, 1, tumor protein p53 binding protein 1, tumor protein p53-binding protein, 1, tumor suppressor p53-binding protein 1 | |
| TP53BP1 | |
| IgG | |
| Affinity Purified | |
| Human, mouse, rat and goat. Human has been tested in WB, IHC-P and ICC/IF, mouse and goat have only been tested in ICC/IF. Primate reactivity reported in scientific literature (PMID: 18389475).Fish reactivity reported in scientific literature (PMID: 25516420). |
| 1 mg/mL | |
| Western Blot, ChIP Assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, In Situ Hybridization (ISH), Immunoassay, KnockDown | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Bat, Bovine, Canine, Fish, Goat, Primate | |
| Q12888 | |
| 7158 | |
| The epitope recognized by 53BP1 Antibody maps to a region between residues 350 and 400 of human 53BP1 [NP_005648.1]. | |
| Primary | |
| Store at -20 degreesC. | |
| 214 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title